Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033241.1 | - | 1 - 2 | 266 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |