Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291208.1 | NP_001278137.1 | 1 - 2 | 446 | 88 | |
GenBank mRNA | AF535870.1 | - | 1 - 2 | 447 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |