Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145949.2 | NP_666061.3 | 7 - 8 | 751 | 88 | |
GenBank mRNA | AK082385.1 | - | 6 - 7 | 942 | 88 | |
BC026393.1 | - | 7 - 8 | 751 | 4195134 | 88 | |
EF137182.1 | - | 7 - 8 | 751 | 88 | ||
EU440733.1 | - | 6 - 7 | 543 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |