Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028083.4 | NP_082359.1 | 8 - 9 | 862 | 70 | |
GenBank mRNA | AK011243.1 | - | 8 - 9 | 863 | 70 | |
BC013532.1 | - | 7 - 8 | 772 | 4238801 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |