Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111027.2 | NP_001104497.1 | 2 - 3 | 616 | 75 | |
GenBank mRNA | BY140482.1 | - | 2 - 3 | 290 | 75 | |
X79989.1 | - | 1 - 2 | 61 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |