Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133756.4 | NP_598517.1 | 9 - 10 | 729 | 61 | |
GenBank mRNA | AK077645.1 | - | 9 - 10 | 730 | 61 | |
AK145957.1 | - | 726 | 61 | |||
AK146686.1 | - | 741 | 61 | |||
BC020174.1 | - | 9 - 10 | 729 | 4505118 | 61 | |
BC096466.1 | - | 9 - 10 | 722 | 3711417 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |