Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008550.2 | NP_032576.2 | 11 - 12 | 1922 | 77 | |
GenBank mRNA | AB006458.1 | - | 11 - 12 | 1900 | 77 | |
AK004900.1 | - | 11 - 12 | 1876 | 77 | ||
AK040388.1 | - | 11 - 12 | 1883 | 77 | ||
AK165376.1 | - | 11 - 12 | 1915 | 77 | ||
BC066211.1 | - | 11 - 12 | 1873 | 30101816 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |