Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008550.2 | NP_032576.2 | 7 - 8 | 1126 | 57 | |
GenBank mRNA | AB006458.1 | - | 7 - 8 | 1104 | 57 | |
AK004900.1 | - | 7 - 8 | 1080 | 57 | ||
AK040388.1 | - | 7 - 8 | 1087 | 57 | ||
AK165376.1 | - | 7 - 8 | 1119 | 57 | ||
BC066211.1 | - | 7 - 8 | 1077 | 30101816 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |