Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011773.3 | NP_035903.2 | 3 - 4 | 598 | 61 | |
GenBank mRNA | AK144568.1 | - | 3 - 4 | 582 | 61 | |
AK165796.1 | - | 3 - 4 | 561 | 61 | ||
BC066199.1 | - | 3 - 4 | 580 | 30476359 | 61 | |
U76007.1 | - | 3 - 4 | 556 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |