Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289757.1 | NP_001276686.1 | 1 - 2 | 116 | 79 | |
NR_110371.1 | 1 - 2 | 116 | 79 | |||
GenBank mRNA | AK012288.1 | - | 1 - 2 | 116 | 79 | |
AK016026.1 | - | 1 - 2 | 59 | 79 | ||
BC061103.1 | - | 1 - 2 | 54 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |