Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK181231.1 | - | 1 - 2 | 61 | 88 | |
AK212996.1 | - | 1 - 2 | 60 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |