Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159887.1 | NP_001153359.1 | 5 - 6 | 430 | 132 | |
NM_001159888.1 | NP_001153360.1 | 5 - 6 | 430 | 132 | ||
NM_001159889.1 | NP_001153361.1 | 5 - 6 | 430 | 132 | ||
NM_023429.4 | NP_075918.1 | 5 - 6 | 430 | 132 | ||
GenBank mRNA | AF295357.1 | - | 5 - 6 | 381 | 132 | |
AK006013.1 | - | 5 - 6 | 422 | 132 | ||
AK007636.1 | - | 5 - 6 | 414 | 132 | ||
AK043603.1 | - | 5 - 6 | 399 | 132 | ||
AK077904.1 | - | 5 - 6 | 400 | 132 | ||
AK078102.1 | - | 5 - 6 | 431 | 132 | ||
AK078909.1 | - | 5 - 6 | 394 | 132 | ||
AK080795.1 | - | 5 - 6 | 422 | 132 | ||
AK082769.1 | - | 5 - 6 | 424 | 132 | ||
AK082781.1 | - | 5 - 6 | 373 | 132 | ||
AK145907.1 | - | 5 - 6 | 432 | 132 | ||
AK159811.1 | - | 5 - 6 | 422 | 132 | ||
AK167320.1 | - | 5 - 6 | 394 | 132 | ||
AK169715.1 | - | 5 - 6 | 412 | 132 | ||
AK177999.1 | - | 3 - 4 | 174 | 132 | ||
AK217055.1 | - | 4 - 5 | 262 | 132 | ||
AY208935.1 | - | 5 - 6 | 329 | 132 | ||
BC006937.1 | - | 5 - 6 | 337 | 2648369 | 132 | |
CX238085.1 | - | 5 - 6 | 378 | 132 | ||
CX238454.1 | - | 5 - 6 | 364 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |