Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145920.3 | NP_666032.1 | 7 - 8 | 950 | 66 | |
GenBank mRNA | AB083066.1 | - | 7 - 8 | 880 | 66 | |
AK044977.1 | - | 7 - 8 | 821 | 66 | ||
AK134298.1 | - | 7 - 8 | 933 | 66 | ||
AK165051.1 | - | 7 - 8 | 835 | 66 | ||
BC037473.1 | - | 7 - 8 | 931 | 4237472 | 66 | |
BC064473.1 | - | 7 - 8 | 941 | 3156380 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |