Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020295.3 | NP_064691.2 | 1 - 2 | 259 | 78 | |
GenBank mRNA | AF190665.1 | - | 1 - 2 | 61 | 78 | |
BC016110.1 | - | 1 - 2 | 99 | 4481088 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |