Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144538.2 | NP_653121.1 | 6 - 7 | 908 | 58 | |
GenBank mRNA | AK004767.1 | - | 5 - 6 | 684 | 58 | |
AK132764.1 | - | 9 - 10 | 1615 | 58 | ||
AK161298.1 | - | 9 - 10 | 1615 | 58 | ||
BC020147.1 | - | 6 - 7 | 873 | 3708973 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |