Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190733.1 | NP_001177662.1 | 1 - 2 | 164 | 71 | |
NM_178896.5 | NP_849227.1 | 1 - 2 | 164 | 71 | ||
GenBank mRNA | AK033559.1 | - | 1 - 2 | 109 | 71 | |
BC060213.1 | - | 1 - 2 | 105 | 71 | ||
BC108346.1 | - | 1 - 2 | 66 | 6310392 | 71 | |
BC137627.1 | - | 1 - 2 | 108 | 8860646 | 71 | |
BC137628.1 | - | 1 - 2 | 108 | 8860647 | 71 | |
BC144794.1 | - | 1 - 2 | 108 | 9053342 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |