Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253737.1 | NP_001240666.1 | 3 - 4 | 221 | 200 | |
NM_001253739.1 | NP_001240668.1 | 3 - 4 | 232 | 200 | ||
NM_001253743.1 | NP_001240672.1 | 4 - 5 | 281 | 200 | ||
NM_001253746.1 | NP_001240675.1 | 3 - 4 | 221 | 137 | ||
NM_013517.4 | NP_038545.1 | 4 - 5 | 362 | 200 | ||
GenBank mRNA | AK156654.1 | - | 4 - 5 | 365 | 200 | |
AK172591.1 | - | 4 - 5 | 365 | 200 | ||
AY069981.1 | - | 3 - 4 | 187 | 137 | ||
BC119164.1 | - | 3 - 4 | 229 | 8733913 | 200 | |
BC119190.1 | - | 3 - 4 | 229 | 8733939 | 200 | |
M34163.1 | - | 4 - 5 | 364 | 200 | ||
M99371.1 | - | 4 - 5 | 354 | 200 | ||
S67072.1 | - | 3 - 4 | 213 | 200 | ||
S67074.1 | - | 3 - 4 | 132 | 200 | ||
X64223.1 | - | 3 - 4 | 221 | 200 | ||
X64224.1 | - | 3 - 4 | 232 | 200 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |