Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039578.3 | NP_001034667.3 | 1 - 2 | 195 | 113 | |
GenBank mRNA | AK045240.1 | - | 1 - 2 | 196 | 113 | |
AK087482.1 | - | 1 - 2 | 196 | 113 | ||
BC042653.1 | - | 1 - 2 | 175 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |