Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039578.3 | NP_001034667.3 | 5 - 6 | 872 | 64 | |
GenBank mRNA | AK045240.1 | - | 5 - 6 | 873 | 64 | |
BC042653.1 | - | 5 - 6 | 852 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |