Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011388.2 | NP_035518.1 | 4 - 5 | 810 | 83 | |
GenBank mRNA | AB002693.1 | - | 4 - 5 | 810 | 83 | |
AY529655.1 | - | 4 - 5 | 810 | 83 | ||
BC120703.1 | - | 4 - 5 | 791 | 40129626 | 83 | |
BC125290.1 | - | 4 - 5 | 791 | 40129805 | 83 | |
D87059.1 | - | 4 - 5 | 737 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |