Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145858.1 | NP_001139330.1 | 10 - 11 | 1628 | 105 | |
NM_001145859.1 | NP_001139331.1 | 10 - 11 | 1631 | 105 | ||
NM_011893.3 | NP_036023.3 | 10 - 11 | 1528 | 105 | ||
GenBank mRNA | AK089455.1 | - | 10 - 11 | 1459 | 105 | |
AK143548.1 | - | 10 - 11 | 1633 | 105 | ||
AK150265.1 | - | 10 - 11 | 1630 | 105 | ||
AK155103.1 | - | 10 - 11 | 1485 | 105 | ||
AK155898.1 | - | 10 - 11 | 1528 | 105 | ||
AK155970.1 | - | 10 - 11 | 1576 | 105 | ||
BC010198.1 | - | 10 - 11 | 1466 | 3967543 | 105 | |
BC085497.1 | - | 10 - 11 | 1519 | 5696296 | 105 | |
L14543.1 | - | 10 - 11 | 1462 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |