Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289803.1 | NP_001276732.1 | 21 - 22 | 2473 | 91 | |
NM_001289804.1 | NP_001276733.1 | 22 - 23 | 2444 | 91 | ||
NM_181857.4 | NP_862905.1 | 22 - 23 | 2527 | 91 | ||
GenBank mRNA | AK133063.1 | - | 22 - 23 | 2364 | 91 | |
AY135562.3 | - | 22 - 23 | 2449 | 91 | ||
BC141101.1 | - | 22 - 23 | 2427 | 9055713 | 91 | |
BC145378.1 | - | 21 - 22 | 2373 | 9053936 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |