Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028499.2 | NP_082775.1 | 2 - 3 | 322 | 120 | |
GenBank mRNA | AK006601.1 | - | 2 - 3 | 333 | 120 | |
AK161371.1 | - | 2 - 3 | 325 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |