Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC029211.1 | - | 5 - 6 | 463 | 4526925 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |