Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111096.1 | NP_001104566.1 | 1 - 2 | 242 | 106 | |
GenBank mRNA | AK151914.1 | - | 1 - 2 | 240 | 106 | |
AK152199.1 | - | 1 - 2 | 240 | 106 | ||
AK156519.1 | - | 1 - 2 | 237 | 106 | ||
AK170561.1 | - | 1 - 2 | 240 | 106 | ||
BC031547.1 | - | 1 - 2 | 212 | 3589207 | 106 | |
BY743506.1 | - | 1 - 2 | 247 | 106 | ||
M57696.1 | - | 1 - 2 | 78 | 106 | ||
M64608.1 | - | 1 - 2 | 228 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |