Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500004283.mm.1 |
TC0500000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500004283.mm.1 |
NM_001289535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004283.mm.1 |
NM_001289536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004283.mm.1 |
NM_026959.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004287.mm.1 |
TC0500000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500004287.mm.1 |
NM_001289535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004287.mm.1 |
NM_001289536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004287.mm.1 |
NM_026959.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004284.mm.1 |
TC0500000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500004284.mm.1 |
NM_001289535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004284.mm.1 |
NM_001289536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500004284.mm.1 |
NM_026959.3 |
Refseq |