Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007937.3 | NP_031963.2 | 14 - 15 | 3242 | 85 | |
GenBank mRNA | BC057401.1 | - | 13 - 14 | 3151 | 5697758 | 85 |
U07357.1 | - | 14 - 15 | 2782 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |