Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem255b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143671.1 | NP_001137143.1 | 4 - 5 | 508 | 66 | |
NM_001143672.1 | NP_001137144.1 | 3 - 4 | 445 | 66 | ||
GenBank mRNA | AK131817.1 | - | 4 - 5 | 508 | 66 | |
BC039763.1 | - | 2 - 3 | 272 | 66 | ||
BC042836.1 | - | 2 - 3 | 261 | 66 | ||
BC089620.1 | - | 3 - 4 | 420 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |