Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081205.1 | NP_001074674.1 | 1 - 2 | 132 | 113 | |
GenBank mRNA | AK075654.1 | - | 1 - 2 | 133 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |