Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173189.2 | NP_775281.2 | 1 - 2 | 467 | 94 | |
GenBank mRNA | AK042026.1 | - | 1 - 2 | 476 | 94 | |
AK083572.1 | - | 1 - 2 | 510 | 94 | ||
AK138858.1 | - | 1 - 2 | 486 | 94 | ||
AK147997.1 | - | 1 - 2 | 59 | 94 | ||
AK148021.1 | - | 1 - 2 | 65 | 94 | ||
BC056924.1 | - | 1 - 2 | 467 | 5697978 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |