Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021315.2 | NP_067290.2 | 16 - 17 | 1911 | 150 | |
GenBank mRNA | AB077991.1 | - | 16 - 17 | 1874 | 150 | |
AF233884.1 | - | 5 - 6 | 374 | 150 | ||
AK134051.1 | - | 16 - 17 | 1870 | 150 | ||
AK168944.1 | - | 16 - 17 | 1914 | 150 | ||
BC031132.1 | - | 16 - 17 | 1874 | 4954951 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |