Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dntt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043228.1 | NP_001036693.1 | 2 - 3 | 538 | 98 | |
NM_009345.2 | NP_033371.2 | 2 - 3 | 538 | 98 | ||
GenBank mRNA | AF316014.1 | - | 2 - 3 | 391 | 98 | |
AF316015.1 | - | 2 - 3 | 391 | 98 | ||
AK039804.1 | - | 2 - 3 | 429 | 98 | ||
AK087978.1 | - | 2 - 3 | 539 | 98 | ||
AK088709.1 | - | 2 - 3 | 539 | 98 | ||
AK134009.1 | - | 2 - 3 | 432 | 98 | ||
X04123.1 | - | 2 - 3 | 425 | 98 | ||
X68670.2 | - | 2 - 3 | 420 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |