Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011037.4 | NP_035167.4 | 3 - 4 | 408 | 81 | |
GenBank mRNA | BC148232.1 | - | 3 - 4 | 412 | 40142573 | 81 |
BC150484.1 | - | 3 - 4 | 412 | 40142579 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |