Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028102.1 | NP_082378.1 | 14 - 15 | 1787 | 71 | |
GenBank mRNA | BC046229.1 | - | 14 - 15 | 1707 | 71 | |
BC118962.2 | - | 12 - 13 | 1780 | 40095026 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |