Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028102.1 | NP_082378.1 | 10 - 11 | 1345 | 130 | |
GenBank mRNA | AK079311.1 | - | 9 - 10 | 1325 | 130 | |
AK173010.1 | - | 8 - 9 | 1020 | 130 | ||
BC046229.1 | - | 9 - 10 | 1265 | 130 | ||
BC118962.2 | - | 10 - 11 | 1581 | 40095026 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |