Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003520.1 | 8 - 9 | 1358 | 136 | ||
GenBank mRNA | AK076430.1 | - | 13 - 14 | 1629 | 136 | |
AK150023.1 | - | 8 - 9 | 1357 | 136 | ||
AK150236.1 | - | 8 - 9 | 1357 | 136 | ||
EU638327.1 | - | 4 - 5 | 682 | 136 | ||
EU638328.1 | - | 7 - 8 | 1175 | 136 | ||
M21039.1 | - | 10 - 11 | 1453 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Probe spans exons |