Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 18 - 19 | 2085 | 68 | |
GenBank mRNA | BC049609.1 | - | 6 - 7 | 586 | 68 | |
BC129920.1 | - | 18 - 19 | 2051 | 68 | ||
X64227.1 | - | 18 - 19 | 2095 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |