Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 7 - 8 | 665 | 97 | |
GenBank mRNA | BC060975.1 | - | 7 - 8 | 628 | 97 | |
BC129920.1 | - | 7 - 8 | 631 | 97 | ||
X64227.1 | - | 7 - 8 | 675 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |