Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028971.1 | NP_083247.1 | 11 - 12 | 1394 | 112 | |
GenBank mRNA | AK016993.1 | - | 11 - 12 | 1322 | 112 | |
AK029938.1 | - | 11 - 12 | 1362 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |