Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039543.2 | NP_001034632.1 | 3 - 4 | 361 | 101 | |
GenBank mRNA | BC027526.1 | - | 3 - 4 | 310 | 1495721 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |