Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138591.2 | NP_613057.2 | 3 - 4 | 489 | 88 | |
GenBank mRNA | AK033214.1 | - | 3 - 4 | 488 | 88 | |
BC013093.1 | - | 3 - 4 | 492 | 3584730 | 88 | |
BC031772.1 | - | 3 - 4 | 456 | 4239980 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |