Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138591.2 | NP_613057.2 | 7 - 8 | 1122 | 111 | |
GenBank mRNA | AF315511.1 | - | 7 - 8 | 1089 | 111 | |
AK033214.1 | - | 7 - 8 | 1121 | 111 | ||
BC013093.1 | - | 7 - 8 | 1125 | 3584730 | 111 | |
BC031772.1 | - | 7 - 8 | 1089 | 4239980 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |