Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163310.1 | NP_001156782.1 | 3 - 4 | 233 | 77 | |
NM_009069.4 | NP_033095.1 | 4 - 5 | 378 | 77 | ||
NR_028056.1 | 4 - 5 | 407 | 77 | |||
GenBank mRNA | AK005357.1 | - | 4 - 5 | 339 | 77 | |
AK014126.1 | - | 4 - 5 | 369 | 77 | ||
AK164196.1 | - | 4 - 5 | 369 | 77 | ||
BC012694.1 | - | 4 - 5 | 338 | 4023732 | 77 | |
CV562852.1 | - | 4 - 5 | 346 | 77 | ||
CX242151.1 | - | 3 - 4 | 181 | 77 | ||
U71205.1 | - | 4 - 5 | 315 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |