Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122820.1 | NP_001116292.1 | 1 - 2 | 135 | 108 | |
GenBank mRNA | AK134634.1 | - | 1 - 2 | 85 | 108 | |
BY121529.1 | - | 1 - 2 | 94 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |