Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC049113.1 | - | 8 - 9 | 1353 | 81 | |
BC057174.1 | - | 8 - 9 | 1103 | 5004450 | 81 | |
BC145338.1 | - | 7 - 8 | 886 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |