Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163493.1 | NP_001156965.1 | 3 - 4 | 426 | 63 | |
NM_146258.2 | NP_666370.3 | 3 - 4 | 426 | 63 | ||
GenBank mRNA | AK050259.1 | - | 3 - 4 | 427 | 63 | |
AK148227.1 | - | 3 - 4 | 385 | 63 | ||
BC027830.1 | - | 3 - 4 | 438 | 5320966 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |