Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lca5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001492.2 | NP_001001492.2 | 5 - 6 | 1148 | 72 | |
GenBank mRNA | AK029554.1 | - | 5 - 6 | 1142 | 72 | |
AK133316.1 | - | 5 - 6 | 1149 | 72 | ||
BC141285.1 | - | 4 - 5 | 998 | 9055897 | 72 | |
BC145611.1 | - | 4 - 5 | 998 | 9054177 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |