Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029440.3 | NP_083716.2 | 16 - 17 | 1460 | 66 | |
GenBank mRNA | AK015309.1 | - | 14 - 15 | 1564 | 66 | |
AK087389.1 | - | 16 - 17 | 1460 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |