Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029440.3 | NP_083716.2 | 18 - 19 | 1662 | 75 | |
GenBank mRNA | AK015309.1 | - | 16 - 17 | 1766 | 75 | |
AK087389.1 | - | 18 - 19 | 1662 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |