Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029440.3 | NP_083716.2 | 7 - 8 | 599 | 88 | |
GenBank mRNA | AK087389.1 | - | 7 - 8 | 599 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |